MilliporeSigma
  • MiR-128 up-regulation inhibits Reelin and DCX expression and reduces neuroblastoma cell motility and invasiveness.
MiR-128 up-regulation inhibits Reelin and DCX expression and reduces neuroblastoma cell motility and invasiveness.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-08-29)
Cristina Evangelisti, Maria Carolina Florian, Isabella Massimi, Carlo Dominici, Giuseppe Giannini, Silvia Galardi, Maria Cristina Buè, Simone Massalini, Heather P McDowell, Elio Messi, Alberto Gulino, Maria Giulia Farace, Silvia Anna Ciafrè
ABSTRACT
MicroRNAs are a class of sophisticated regulators of gene expression, acting as post-transcriptional inhibitors that recognize their target mRNAs through base pairing with short regions along the 3'UTRs. Several microRNAs are tissue specific, suggesting a specialized role in tissue differentiation or maintenance, and quite a few are critically involved in tumorigenesis. We studied miR-128, a brain-enriched microRNA, in retinoic acid-differentiated neuroblastoma cells, and we found that this microRNA is up-regulated in treated cells, where it down-modulates the expression of two proteins involved in the migratory potential of neural cells: Reelin and DCX. Consistently, miR-128 ectopic overexpression suppressed Reelin and DCX, whereas the LNA antisense-mediated miR-128 knockdown caused the two proteins to increase. Ectopic miR-128 overexpression reduced neuroblastoma cell motility and invasiveness, and impaired cell growth. Finally, the analysis of a small series of primary human neuroblastomas showed an association between high levels of miR-128 expression and favorable features, such as favorable Shimada category or very young age at diagnosis. Thus, we provide evidence for a role for miR-128 in the molecular events modulating neuroblastoma progression and aggressiveness.
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, HRP conjugate, Chemicon®, from rabbit