MilliporeSigma
The controlled evolution of a polymer single crystal.
Science (New York, N.Y.) (2005-03-19)
Xiaogang Liu, Yi Zhang, Dipak K Goswami, John S Okasinski, Khalid Salaita, Peng Sun, Michael J Bedzyk, Chad A Mirkin
ABSTRACT
We present a method for controlling the initiation and kinetics of polymer crystal growth using dip-pen nanolithography and an atomic force microscope tip coated with poly-dl-lysine hydrobromide. Triangular prisms of the polymer epitaxially grow on freshly cleaved mica substrates, and their in-plane and out-of-plane growth rates can be controlled by raster scanning the coated tip across the substrate. Atomic force microscope images were concomitantly recorded, providing a set of photographic images of the process as it spans the nanometer- to micrometer-length scales as a function of environmental conditions.
Sigma-Aldrich
Poly-DL-lysine hydrobromide, mol wt >40,000
Sigma-Aldrich
Poly-DL-lysine hydrobromide, mol wt 25,000-40,000