MilliporeSigma
  • Effect of hypoxia on integrin-mediated adhesion of endothelial progenitor cells.
Effect of hypoxia on integrin-mediated adhesion of endothelial progenitor cells.
Journal of cellular and molecular medicine (2012-02-23)
Ralf Kaiser, Denise Friedrich, Emmanouil Chavakis, Michael Böhm, Erik B Friedrich
ABSTRACT
Homing of endothelial progenitor cells (EPCs) is crucial for neoangiogenesis, which might be negatively affected by hypoxia. We investigated the influence of hypoxia on fibronectin binding integrins for migration and cell-matrix-adhesion. AMP-activated kinase (AMPK) and integrin-linked kinase (ILK) were examined as possible effectors of hypoxia. Human EPCs were expanded on fibronectin (FN) and integrin expression was profiled by flow cytometry. Cell-matrix-adhesion- and migration-assays on FN were performed to examine the influence of hypoxia and AMPK-activation. Regulation of AMPK and ILK was shown by Western blot analysis. We demonstrate the presence of integrin β(1), β(2) and α(5) on EPCs. Adhesion to FN is reduced by blocking β(1) and α(5) (49% and 2% of control, P < 0.05) whereas α(4)-blockade has no effect. Corresponding effects were shown for migration. Hypoxia and AMPK-activation decrease adhesion on FN. Although total AMPK-expression remains unchanged, phospho-AMPK increases eightfold. The EPCs require α(5) for adhesion on FN. Hypoxia and AMPK-activation decrease adhesion. As α(5) is the major adhesive factor for EPCs on FN, this suggests a link between AMPK and α(5)-integrins. We found novel evidence for a connection between hypoxia, AMPK-activity and integrin activity. This might affect the fate of EPCs in ischaemic tissue.
Sigma-Aldrich
Anti-Integrin β2 [CD18] Antibody, clone MEM 48, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, Azide Free, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α5 Antibody, clone P1D6, azide free, clone P1D6, Chemicon®, from mouse
Millipore
Anti-Integrin β1 Antibody, clone TDM29, clone TDM29, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, azide free, clone P4G11, Chemicon®, from mouse