MilliporeSigma
  • Expression and characterization of full-length human huntingtin, an elongated HEAT repeat protein.
Expression and characterization of full-length human huntingtin, an elongated HEAT repeat protein.
The Journal of biological chemistry (2006-04-06)
Wei Li, Louise C Serpell, Wendy J Carter, David C Rubinsztein, James A Huntington
ABSTRACT
Huntington disease is an inherited neurodegenerative disorder that is caused by expanded CAG trinucleotide repeats, resulting in a polyglutamine stretch of >37 on the N terminus of the protein huntingtin (htt). htt is a large (347 kDa), ubiquitously expressed protein. The precise functions of htt are not clear, but its importance is underscored by the embryonic lethal phenotype in htt knock-out mice. Despite the fact that the htt gene was cloned 13 years ago, little is known about the properties of the full-length protein. Here we report the expression and preliminary characterization of recombinant full-length wild-type human htt. Our results support a model of htt composed entirely of HEAT repeats that stack to form an elongated superhelix.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 2146-2541, clone HU-2E8, ascites fluid, clone HU-2E8, Chemicon®
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 1247-1646, clone HU-4E6, ascites fluid, clone HU-4E6, Chemicon®