MilliporeSigma
  • The coupling of 5-oxo-eicosanoid receptors to heterotrimeric G proteins.
The coupling of 5-oxo-eicosanoid receptors to heterotrimeric G proteins.
Journal of immunology (Baltimore, Md. : 1950) (2000-03-08)
J T O'Flaherty, J S Taylor, M Kuroki
ABSTRACT
5-Oxo-eicosatetraenoic acid (5-oxoETE) stimulated human neutrophil (PMN) and eosinophil chemotaxis, PMN hexose uptake, and PMN membrane GTP/GDP exchange. Pertussis toxin (PT), a blocker of heterotrimeric G proteins (GP), completely inhibited these responses, but proved far less effective on the same responses when elicited by leukotriene B4, C5a, FMLP, platelet-activating factor, IL-8, or RANTES chemotactic factors. 5-OxoETE also specifically bound to the membrane preparations that conducted GTP/GDP exchange. This binding was down-regulated by GTPgammaS, but not ADPgammaS, and displaced by 5-oxoETE analogues, but not by leukotriene B4, lipoxin A4, or lipoxin B4. Finally, PMN expressed PT-sensitive GP alphaiota2 and PT-resistant GP alphaq/11- and alpha13-chains; eosinophils expressed only alphai2 and alphaq/11. We conclude that 5-oxoETE activates granulocytes through a unique receptor that couples preferentially to PT-sensitive GP. The strict dependency of this putative receptor on PT-sensitive GP may underlie the limited actions of 5-oxoETE, compared with other CF, and help clarify the complex relations between receptors, GP, cell signals, and cell responses.
Sigma-Aldrich
Anti-G Protein Giα-2 Antibody, clone L5, clone L5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Giα-3-Subunit, C-Terminal (345-354) Rabbit pAb, liquid, Calbiochem®