MilliporeSigma
  • Suzuki-Miyaura cross-coupling reaction on copper-trans-A(2)B corroles with excellent functional group tolerance.
Suzuki-Miyaura cross-coupling reaction on copper-trans-A(2)B corroles with excellent functional group tolerance.
Tetrahedron (2011-07-16)
Michael König, Lorenz Michael Reith, Uwe Monkowius, Günther Knör, Klaus Bretterbauer, Wolfgang Schoefberger
ABSTRACT
The palladium-catalyzed Suzuki-Miyaura cross-coupling reaction has been investigated on meso-substituted trans-A(2)B-corrole using tailored Pd-catalyst systems.We present the first examples of Suzuki-Miyaura cross-coupling reactions on meso-substituted trans-A(2)B-corrole derivatives with neutral, sterically hindered, inactivated and heteroaromatic boronic acids and esters, alkenylboronic acids, as well as quickly deboronating aryl boronic acids and benzo-condensated five membered heterocyclic boronic acids. In addition, we established a high-yield procedure for the Suzuki-Miyaura cross-coupling reaction of corroles with neutral boronic acids.Due to the lability of the free-base corrole macrocycles, functionalization of the corrole periphery was performed with the corresponding Cu-metallated species. meso-Substituted trans-A(2)B-corrole can hence be regarded as highly versatile platform towards more sophisticated corrole systems.X-ray structure analysis of a functionalized meso-substituted trans-A(2)B copper corrole exhibited the typical features of such a Cu-complex: short N-Cu distances and a saddled corrole configuration.Moreover, we observed a sensitivity of the formal oxidation state of the coordinated copper ions towards Suzuki-Miyaura cross-coupling reaction conditions, where the central copper(III) ion approaches the characteristic features of a copper(II) species. This redox behaviour was examined by UV/vis absorption spectra, nuclear magnetic resonance (NMR) experiments and time-dependent density functional theoretical calculations.
Sigma-Aldrich
4-Aminophenylboronic acid pinacol ester, 97%