MilliporeSigma
  • Asymmetric Iron-Catalyzed C-H Alkylation Enabled by Remote Ligand meta-Substitution.
Asymmetric Iron-Catalyzed C-H Alkylation Enabled by Remote Ligand meta-Substitution.
Angewandte Chemie (International ed. in English) (2017-09-19)
Joachim Loup, Daniel Zell, João C A Oliveira, Helena Keil, Dietmar Stalke, Lutz Ackermann
ABSTRACT
Highly enantioselective iron-catalyzed C-H alkylations by inner-sphere C-H activation were accomplished with ample scope. High levels of enantiocontrol proved viable through a novel ligand design that exploits a remote meta-substitution on N-heterocyclic carbenes within a facile ligand-to-ligand H-transfer C-H cleavage.
Sigma-Aldrich
Vinylferrocene, 97%