MilliporeSigma
  • Ploidy-dependent change in cyclin D2 expression and sensitization to cdk4/6 inhibition in human somatic haploid cells.
Ploidy-dependent change in cyclin D2 expression and sensitization to cdk4/6 inhibition in human somatic haploid cells.
Biochemical and biophysical research communications (2018-09-09)
Kan Yaguchi, Takahiro Yamamoto, Masaya Shimada, Rina Sugimoto, Kiminori Nakamura, Tokiyoshi Ayabe, Ryota Uehara
ABSTRACT
Near-haploidy is observed in certain cancer types, but ploidy-dependent alterations in gene regulation in the haploid state remain elusive. Here, by comparative transcriptome analysis between human isogenic haploid and diploid cell lines, we found lowering of cyclin D2 level in haploids. Acute genome duplication in haploids restored cyclin D2 expression to diploid level, indicating that the regulation of cyclin D2 expression is directly linked to ploidy. Downstream pathways of cyclin D2, such as Rb phosphorylation and p27 sequestration remained intact in haploids, suggesting that they adapt to lowered cyclin D level. Interestingly, however, haploid cells were more susceptible to cdk4/6 inhibition compared to diploids. Our finding indicates feasibility of selective growth suppression of haploid cells based on ploidy-linked gene regulation.
Sigma-Aldrich
PD 0332991 isethionate, ≥98% (HPLC)
Pricing and availability is not currently available.