MilliporeSigma
  • Surface chemistry of gold nanoparticles determines the biocorona composition impacting cellular uptake, toxicity and gene expression profiles in human endothelial cells.
Surface chemistry of gold nanoparticles determines the biocorona composition impacting cellular uptake, toxicity and gene expression profiles in human endothelial cells.
Nanotoxicology (2017-04-20)
Parwathy Chandran, Jim E Riviere, Nancy A Monteiro-Riviere
ABSTRACT
This study investigated the role of nanoparticle size and surface chemistry on biocorona composition and its effect on uptake, toxicity and cellular responses in human umbilical vein endothelial cells (HUVEC), employing 40 and 80 nm gold nanoparticles (AuNP) with branched polyethyleneimine (BPEI), lipoic acid (LA) and polyethylene glycol (PEG) coatings. Proteomic analysis identified 59 hard corona proteins among the various AuNP, revealing largely surface chemistry-dependent signature adsorbomes exhibiting human serum albumin (HSA) abundance. Size distribution analysis revealed the relative instability and aggregation inducing potential of bare and corona-bound BPEI-AuNP, over LA- and PEG-AuNP. Circular dichroism analysis showed surface chemistry-dependent conformational changes of proteins binding to AuNP. Time-dependent uptake of bare, plasma corona (PC) and HSA corona-bound AuNP (HSA-AuNP) showed significant reduction in uptake with PC formation. Cell viability studies demonstrated dose-dependent toxicity of BPEI-AuNP. Transcriptional profiling studies revealed 126 genes, from 13 biological pathways, to be differentially regulated by 40 nm bare and PC-bound BPEI-AuNP (PC-BPEI-AuNP). Furthermore, PC formation relieved the toxicity of cationic BPEI-AuNP by modulating expression of genes involved in DNA damage and repair, heat shock response, mitochondrial energy metabolism, oxidative stress and antioxidant response, and ER stress and unfolded protein response cascades, which were aberrantly expressed in bare BPEI-AuNP-treated cells. NP surface chemistry is shown to play the dominant role over size in determining the biocorona composition, which in turn modulates cell uptake, and biological responses, consequently defining the potential safety and efficacy of nanoformulations.
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 10nm diameter, Cy3 and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and maleimide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and methyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and protein A functionalized, powder