MilliporeSigma
Unraveling the long-pursued Au144 structure by x-ray crystallography.
Science advances (2018-10-20)
Nan Yan, Nan Xia, Lingwen Liao, Min Zhu, Fengming Jin, Rongchao Jin, Zhikun Wu
ABSTRACT
The transition from nanocluster to nanocrystal is a central issue in nanoscience. The atomic structure determination of metal nanoparticles in the transition size range is challenging and particularly important in understanding the quantum size effect at the atomic level. On the basis of the rationale that the intra- and interparticle weak interactions play critical roles in growing high-quality single crystals of metal nanoparticles, we have reproducibly obtained ideal crystals of Au144(SR)60 and successfully solved its structure by x-ray crystallography (XRC); this structure was theoretically predicted a decade ago and has long been pursued experimentally but without success until now. Here, XRC reveals an interesting Au12 hollow icosahedron in thiolated gold nanoclusters for the first time. The Au-Au bond length, close to that of bulk gold, shows better thermal extensibility than the other Au-Au bond lengths in Au144(SR)60, providing an atomic-level perspective because metal generally shows better thermal extensibility than nonmetal materials. Thus, our work not only reveals the mysterious, long experimentally pursued structure of a transition-sized nanoparticle but also has important implications for the growth of high-quality, single-crystal nanoparticles, as well as for the understanding of the thermal extensibility of metals from the perspective of chemical bonding.
Sigma-Aldrich
4-tert-Butylbenzyl mercaptan
Sigma-Aldrich
2,4-Dimethylbenzenethiol, 95%
Sigma-Aldrich
1-Phenylethyl mercaptan, AldrichCPR