MilliporeSigma
  • Subversion of Systemic Glucose Metabolism as a Mechanism to Support the Growth of Leukemia Cells.
Subversion of Systemic Glucose Metabolism as a Mechanism to Support the Growth of Leukemia Cells.
Cancer cell (2018-10-03)
Haobin Ye, Biniam Adane, Nabilah Khan, Erica Alexeev, Nichole Nusbacher, Mohammad Minhajuddin, Brett M Stevens, Amanda C Winters, Xi Lin, John M Ashton, Enkhtsetseg Purev, Lianping Xing, Daniel A Pollyea, Catherine A Lozupone, Natalie J Serkova, Sean P Colgan, Craig T Jordan
ABSTRACT
From an organismal perspective, cancer cell populations can be considered analogous to parasites that compete with the host for essential systemic resources such as glucose. Here, we employed leukemia models and human leukemia samples to document a form of adaptive homeostasis, where malignant cells alter systemic physiology through impairment of both host insulin sensitivity and insulin secretion to provide tumors with increased glucose. Mechanistically, tumor cells induce high-level production of IGFBP1 from adipose tissue to mediate insulin sensitivity. Further, leukemia-induced gut dysbiosis, serotonin loss, and incretin inactivation combine to suppress insulin secretion. Importantly, attenuated disease progression and prolonged survival are achieved through disruption of the leukemia-induced adaptive homeostasis. Our studies provide a paradigm for systemic management of leukemic disease.
Sigma-Aldrich
Barium hydroxide solution, 0.3 N
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 4,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Carmine, powder, ≥42.0% (UV-vis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Supelco
Serotonin, analytical standard
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture