MilliporeSigma
  • Multiclass screening method to detect more than fifty banned substances in bovine bile and urine.
Multiclass screening method to detect more than fifty banned substances in bovine bile and urine.
Analytica chimica acta (2018-08-26)
Simone Moretti, Francesca Lega, Lorenzo Rigoni, Giorgio Saluti, Danilo Giusepponi, Antimo Gioiello, Elisabetta Manuali, Rosanna Rossi, Roberta Galarini
ABSTRACT
A multiclass screening method to detect fifty-three forbidden substances by liquid-chromatography coupled to hybrid high-resolution mass spectrometry (LC-Q-Orbitrap) was developed and validated in bovine bile and urine. Eight classes of compounds were included in the method's scope (β-agonists, corticosteroids, nitroimidazoles, progestins, resorcylic acid lactones (RALs), sedatives, steroids and stilbenes) plus chloramphenicol and dapsone. After hydrolysis, the sample was divided in two aliquots, which followed two parallel purification steps. The reunified extracts were injected and two chromatographic runs performed in positive and negative ionization mode, respectively. The validation data (60 different samples per matrix) proved that the method was fit for purpose with detection capabilities lower than 1 μg L-1 in both matrices. The combined application of accurate mass acquisition and two-stage mass spectrometry (parallel reaction monitoring) was crucial to achieve suitable selectivity, which is the most critical parameter mainly for urines. Finally, the long-standing problem of the high rate of false positive results for RALs, due to the natural ingestion of mycotoxin, zearalenone, was taken on including all their labelled standards. That allowed a very satisfactory management of this screening test.
Sigma-Aldrich
Beclomethasone, ≥99%
Sigma-Aldrich
Flumethasone
Supelco
HMMNI, VETRANAL®, analytical standard
Supelco
Ipronidazole-OH, VETRANAL®, analytical standard
Supelco
Carazolol, analytical standard
Supelco
Ronidazole, analytical standard
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Supelco
Metronidazole-OH, VETRANAL®, analytical standard
Supelco
Ipronidazole, VETRANAL®, analytical standard