MilliporeSigma
  • Commensal microflora-induced T cell responses mediate progressive neurodegeneration in glaucoma.
Commensal microflora-induced T cell responses mediate progressive neurodegeneration in glaucoma.
Nature communications (2018-08-12)
Huihui Chen, Kin-Sang Cho, T H Khanh Vu, Ching-Hung Shen, Mandeep Kaur, Guochun Chen, Rose Mathew, M Lisa McHam, Ahad Fazelat, Kameran Lashkari, Ngan Pan Bennett Au, Joyce Ka Yu Tse, Yingqian Li, Honghua Yu, Lanbo Yang, Joan Stein-Streilein, Chi Him Eddie Ma, Clifford J Woolf, Mark T Whary, Martine J Jager, James G Fox, Jianzhu Chen, Dong F Chen
ABSTRACT
Glaucoma is the most prevalent neurodegenerative disease and a leading cause of blindness worldwide. The mechanisms causing glaucomatous neurodegeneration are not fully understood. Here we show, using mice deficient in T and/or B cells and adoptive cell transfer, that transient elevation of intraocular pressure (IOP) is sufficient to induce T-cell infiltration into the retina. This T-cell infiltration leads to a prolonged phase of retinal ganglion cell degeneration that persists after IOP returns to a normal level. Heat shock proteins (HSP) are identified as target antigens of T-cell responses in glaucomatous mice and human glaucoma patients. Furthermore, retina-infiltrating T cells cross-react with human and bacterial HSPs; mice raised in the absence of commensal microflora do not develop glaucomatous T-cell responses or the associated neurodegeneration. These results provide compelling evidence that glaucomatous neurodegeneration is mediated in part by T cells that are pre-sensitized by exposure to commensal microflora.
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder