MilliporeSigma
  • Novel β-phenylacrylic acid derivatives exert anti-cancer activity by inducing Src-mediated apoptosis in wild-type KRAS colon cancer.
Novel β-phenylacrylic acid derivatives exert anti-cancer activity by inducing Src-mediated apoptosis in wild-type KRAS colon cancer.
Cell death & disease (2018-08-31)
Su Jin Kim, Tae Hwan Noh, Sujin Son, Do Hyun Kim, Wooseong Kim, Yunna Lee, Jieun Choo, Gwangbeom Heo, Min Jae Kim, Hae Young Chung, Yunjin Jung, Jee Hyung Jung, Hyung Ryong Moon, Eunok Im
ABSTRACT
Many stress conditions including chemotherapy treatment is known to activate Src and under certain condition Src can induce the apoptotic signal via c-Jun N-terminal kinase (JNK) activation. Here we report that the newly synthesized β-phenylacrylic acid derivatives, MHY791 and MHY1036 (MHYs), bind to epidermal growth factor receptor (EGFR) tyrosine kinase domains and function as EGFR inhibitors, having anti-cancer activities selectively in wild-type KRAS colon cancer. Mechanistically, MHYs-induced Src/JNK activation which enhanced their pro-apoptotic effects and therefore inhibition of Src by the chemical inhibitor PP2 or Src siRNA abolished the response. In addition, MHYs generated reactive oxygen species and increased ER stress, and pretreatment with antioxidant-inhibited MHY-induced ER stress, Src activation, and apoptosis. Furthermore, the irreversible EGFR inhibitor PD168393 also activated Src while the reversible EGFR inhibitor gefitinib showed the opposite effect, indicating that MHYs are the irreversible EGFR inhibitor. Collectively, Src can play a key role in apoptosis induced by the novel EGFR inhibitor MHYs, suggesting that activation of Src might prove effective in treating EGFR/wild-type KRAS colon cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
PD168393, ≥98% (HPLC)