MilliporeSigma
  • Reflectance variation in the blue tit crown in relation to feather structure.
Reflectance variation in the blue tit crown in relation to feather structure.
The Journal of experimental biology (2018-04-05)
Gergely Hegyi, Miklós Laczi, Dóra Kötél, Tamás Csizmadia, Péter Lőw, Balázs Rosivall, Eszter Szöllősi, János Török
ABSTRACT
Structural plumage colour is one of the most enigmatic sexually selected traits. The information content of structural colour variation is debated, and the heterogeneity of the findings is hard to explain because the proximate background of within-species colour differences is very scarcely studied. We combined measurements of feather macrostructure and nanostructure to explain within-population variability in blue tit crown reflectance. We found that sexual dichromatism in aspects of crown reflectance was explained only by feather macrostructure, whereas nanostructural predictors accounted for some of the age-related differences in reflectance. Moreover, we found that both mean reflectance and spectral shape traits reflected a combination of quantity and regularity aspects in macrostructure and nanostructure. This rich proximate background provides ample scope for reflectance to convey various types of information on individual quality.
Sigma-Aldrich
Spurr Low Viscosity Embedding Kit, hydrophobic resin for embedding histological samples