MilliporeSigma
  • Determination of volatile fatty acids in digestate by solvent extraction with dimethyl carbonate and gas chromatography-mass spectrometry.
Determination of volatile fatty acids in digestate by solvent extraction with dimethyl carbonate and gas chromatography-mass spectrometry.
Analytica chimica acta (2018-09-09)
Michele Ghidotti, Daniele Fabbri, Cristian Torri, Sergio Piccinini
ABSTRACT
Volatile fatty acids (VFAs) are among the most important parameters in process monitoring of anaerobic digestion plants for biogas production. The concentration of single VFA species is typically determined by direct injection of the acidified aqueous phase of digestate samples into GC-FID. Analysis of dimethyl carbonate extracts was investigated as an alternative method consisting of a simple and rapid in-vial procedure of acidification and solvent extraction of the sample, followed by centrifugation and GC-MS analysis. The principal figures of merit resulting from internal standard calibration were comparable to those proposed for the direct analysis of aqueous digestate, while the analysis of real samples did not provide statistically significant differences between the two methods according to parametric and non-parametric tests. Procedural aspects including sample amount and solid removal improved with dimethyl carbonate, while GC contamination was reduced. The method was applied to seventeen samples from fully operating anaerobic digesters fed with various feedstocks and enabled the individuation of high probability of system stress through the values of total VFA, propanoic acid, longer chained VFA concentrations and the ratio between acetic and propanoic acid concentrations. The use of dimethyl carbonate allowed the detection of alicyclic and aromatic acids that could represent new molecular markers in assessing the origin of feed and process conditions.
Sigma-Aldrich
Potassium bisulfate, ≥99.99% trace metals basis