MilliporeSigma
  • Identification of the binding modes of N-phenylphthalimides inhibiting bacterial thymidylate synthase through X-ray crystallography screening.
Identification of the binding modes of N-phenylphthalimides inhibiting bacterial thymidylate synthase through X-ray crystallography screening.
Journal of medicinal chemistry (2011-06-24)
Stefano Mangani, Laura Cancian, Rosalida Leone, Cecilia Pozzi, Sandra Lazzari, Rosaria Luciani, Stefania Ferrari, M Paola Costi
ABSTRACT
To identify specific bacterial thymidylate synthase (TS) inhibitors, we exploited phenolphthalein (PTH), which inhibits both bacterial and human enzymes. The X-ray crystal structure of Lactobacillus casei TS (LcTS) that binds PTH showed multiple binding modes of the inhibitor, which prevented a classical structure-based drug design approach. To overcome this issue, we synthesized two phthalimidic libraries that were tested against TS enzymes and then we performed X-ray crystallographic screening of the active compounds. Compounds 6A, 8A, and 12A showed 40-fold higher affinity for bacterial TS than human TS. The X-ray crystallographic screening characterized the binding mode of six inhibitors in complexes with LcTS. Of these, 20A, 23A, and 24A showed a common unique binding mode, whereas 8A showed a different, unique binding mode. A comparative analysis of the LcTS X-ray complexes that were obtained with the pathogenic TS enabled the selection of compounds 8A and 23A as specific compounds and starting points to be exploited for the specific inhibition of pathogen enzymes.
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Phenolphthalein solution, 1 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein, SAJ first grade, ≥97.0%
Sigma-Aldrich
Phenolphthalein, JIS special grade, ≥98.0%
Sigma-Aldrich
Phenolphthalein solution, 0.04 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein solution, 0.1 % (w/v), pH 7.8-10.0