MilliporeSigma
New approach: Chemical and fluorescence profiling of NZ honeys.
Food chemistry (2018-06-24)
Jessie Bong, Kerry M Loomes, Bin Lin, Jonathan M Stephens
ABSTRACT
New Zealand manuka (Leptospermum scoparium) and kanuka (Kunzea ericoides) honeys contain a unique array of chemical markers useful for chemical fingerprinting. We investigated the presence of 13 potential marker compounds in nectars of the major honey crop species. We confirmed that leptosperin, lepteridine, 2'-methoxyacetophenone, and 2-methoxybenzoic acid are exclusive to manuka nectar whereas lumichrome is unique to kanuka nectar. 3-Phenyllactic acid and 4-hydroxyphenyllactic acid are present in manuka and kanuka nectars. Leptosperin, lepteridine, 3-phenyllactic acid, and 4-hydroxyphenyllactic acid are chemically stable over prolonged storage, but not 2-methoxybenzoic acid and 2'-methoxyacetophenone. Accordingly, leptosperin and lepteridine are definitive chemical markers for authentication of manuka honey. An optimal concentration cut-off was established for the floral source-specific markers: leptosperin (94mg/kg), lepteridine (2.1mg/kg), 2'-methoxyacetophenone (2.0mg/kg) for manuka honey, and lumichrome (4.5mg/kg) for kanuka honey. The use of leptosperin and lepteridine as fluorescence markers for manuka honey authentication is reinforced.
Sigma-Aldrich
Lumichrome
Sigma-Aldrich
3-Phenyllactic acid, ≥98%