MilliporeSigma
  • Knockdown of peroxiredoxin V increases glutamate‑induced apoptosis in HT22 hippocampal neuron cells.
Knockdown of peroxiredoxin V increases glutamate‑induced apoptosis in HT22 hippocampal neuron cells.
Molecular medicine reports (2018-04-06)
Gui-Nan Shen, Lei Liu, Li Feng, Yu Jin, Mei-Hua Jin, Ying-Hao Han, Cheng-Hao Jin, Yong-Zhe Jin, Dong-Soek Lee, Tae Ho Kwon, Yu-Dong Cui, Hu-Nan Sun
ABSTRACT
High concentrations of glutamate may mediate neuronal cell apoptosis by increasing intracellular reactive oxygen species (ROS) levels. Peroxiredoxin V (Prx V), a member of the Prx family, serves crucial roles in protecting cells from oxidative stress. The present study investigated the regulatory effect of Prx V on glutamate‑induced effects on viability and apoptosis in HT22 cells. Western blotting was used for protein expression analysis and Annexin V/PI staining and flow cytometry for determination of apoptosis. The results demonstrated that glutamate may ROS‑dependently increase HT22 cell apoptosis and upregulate Prx V protein levels. Furthermore, knockdown of Prx V protein expression with a lentivirus significantly enhanced HT22 cell apoptosis mediated by glutamate, which was reversed by inhibition of ROS with N‑acetyl‑L‑cysteine. Inhibiting the extracellular signal‑regulated kinase (ERK) signaling pathway with PD98059, a specific inhibitor for ERK phosphorylation, markedly decreased glutamate‑induced HT22 cell apoptosis in Prx V knockdown cells, indicating the potential involvement of ERK signaling in glutamate‑induced HT22 cell apoptosis. In addition, an increase in nuclear apoptosis‑inducing factor was observed in Prx V knockdown HT22 cells following glutamate treatment, compared with mock cells, whereas no differences in B‑cell lymphoma‑2 and cleaved‑caspase‑3 protein expression levels were observed between mock and Prx V knockdown cells. The results of the present study indicated that Prx V may have potential as a therapeutic molecular target for glutamate‑induced neuronal cell death and provide novel insight into the role of Prx V in oxidative‑stress induced neuronal cell death.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
MISSION® esiRNA, targeting human PRDX5
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
Sigma-Aldrich
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder