MilliporeSigma
Defensins of vertebrate animals.
Current opinion in immunology (2002-01-16)
Robert I Lehrer, Tomas Ganz
ABSTRACT
During the past year, novel beta-defensins of mice and men have been identified, together with a novel defensin subfamily (the circular or 'theta' minidefensins) in primates. Insight into the evolution of defensins has been obtained from structural studies, and several mechanisms related to microbial resistance to defensins have been delineated. There is now convincing evidence that defensins augment adaptive immune responses.
Sigma-Aldrich
Beta D-1 (47 aa) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Beta D-1 (36 aa) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Beta D-4 human, recombinant, expressed in E. coli, ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
NP-1 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture