MilliporeSigma
  • Strains of the toxic and bloom-forming Nodularia spumigena (cyanobacteria) can degrade methylphosphonate and release methane.
Strains of the toxic and bloom-forming Nodularia spumigena (cyanobacteria) can degrade methylphosphonate and release methane.
The ISME journal (2018-02-16)
Jonna E Teikari, David P Fewer, Rashmi Shrestha, Shengwei Hou, Niina Leikoski, Minna Mäkelä, Asko Simojoki, Wolfgang R Hess, Kaarina Sivonen
ABSTRACT
Nodularia spumigena is a nitrogen-fixing cyanobacterium that forms toxic blooms in the Baltic Sea each summer and the availability of phosphorous is an important factor limiting the formation of these blooms. Bioinformatic analysis identified a phosphonate degrading (phn) gene cluster in the genome of N. spumigena suggesting that this bacterium may use phosphonates as a phosphorus source. Our results show that strains of N. spumigena could grow in medium containing methylphosphonic acid (MPn) as the sole source of phosphorous and released methane when growing in medium containing MPn. We analyzed the total transcriptomes of N. spumigena UHCC 0039 grown using MPn and compared them with cultures growing in Pi-replete medium. The phnJ, phosphonate lyase gene, was upregulated when MPn was the sole source of phosphorus, suggesting that the expression of this gene could be used to indicate the presence of bioavailable phosphonates. Otherwise, growth on MPn resulted in only a minor reconstruction of the transcriptome and enabled good growth. However, N. spumigena strains were not able to utilize any of the anthropogenic phosphonates tested. The phosphonate utilizing pathway may offer N. spumigena a competitive advantage in the Pi-limited cyanobacterial blooms of the Baltic Sea.
Sigma-Aldrich
Nitrilotri(methylphosphonic acid), ≥95% (T)
Sigma-Aldrich
Ethylphosphonic acid, 98%