MilliporeSigma
  • A Printed Organic Circuit System for Wearable Amperometric Electrochemical Sensors.
A Printed Organic Circuit System for Wearable Amperometric Electrochemical Sensors.
Scientific reports (2018-04-25)
Rei Shiwaku, Hiroyuki Matsui, Kuniaki Nagamine, Mayu Uematsu, Taisei Mano, Yuki Maruyama, Ayako Nomura, Kazuhiko Tsuchiya, Kazuma Hayasaka, Yasunori Takeda, Takashi Fukuda, Daisuke Kumaki, Shizuo Tokito
ABSTRACT
Wearable sensor device technologies, which enable continuous monitoring of biological information from the human body, are promising in the fields of sports, healthcare, and medical applications. Further thinness, light weight, flexibility and low-cost are significant requirements for making the devices attachable onto human tissues or clothes like a patch. Here we demonstrate a flexible and printed circuit system consisting of an enzyme-based amperometric sensor, feedback control and amplification circuits based on organic thin-film transistors. The feedback control and amplification circuits based on pseudo-CMOS inverters were successfuly integrated by printing methods on a plastic film. This simple system worked very well like a potentiostat for electrochemical measurements, and enabled the quantitative and real-time measurement of lactate concentration with high sensitivity of 1 V/mM and a short response time of a hundred seconds.
Sigma-Aldrich
2,3,4,5,6-Pentafluorothiophenol, 97%