MilliporeSigma
  • Structural elucidation and antioxidant activity of an arabinogalactan from the leaves of Moringa oleifera.
Structural elucidation and antioxidant activity of an arabinogalactan from the leaves of Moringa oleifera.
International journal of biological macromolecules (2018-01-26)
Tao-Bin He, Yan-Ping Huang, YeWei Huang, Xuan-Jun Wang, Jiang-Miao Hu, Jun Sheng
ABSTRACT
A novel arabinogalactan (MOP-1) was isolated from leaves of Moringa oleifera and was purified by macro-porous resin, DEAE-52 Cellulose and Sepharose 6B gel filtration chromatography. High performance gel permeation chromatography (HPGPC) analysis showed that the molecular weight of MOP-1 was 7.65 × 107 Da. Monosaccharide composition analysis indicated that sugar composition is Rha: Ara: Gal in a molar ratio of 1:7.32:12.12. Structural analysis indicated that MOP-1 has a backbone of →1)-β-d-Galp-(3,4→ with highly branched chains at O-4 position. The branches were composed of →1)-β-d-Galp-(4→, →1)-α-d-Galp-(2→, Araf-(1→, Galp-(1→. Its antioxidant activities were evaluated by DPPH radical scavenging capacity, 2,2'-Azinobis-(3-ethylbenzthiazoline-6-sulphonate) (ABTS) radical cation scavenging capacity, and ferric-reducing antioxidant power (FRAP). The results indicated MOP-1 exhibited significant antioxidant activities.
Product Description
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000