MilliporeSigma
Photometric detection in flow analysis systems using integrated PEDDs.
Talanta (2008-10-31)
Martina O' Toole, King Tong Lau, Dermot Diamond
ABSTRACT
A novel inexpensive optical-sensing technique has been developed for colorimetric flow analysis. This sensing system employs two LEDs whereby one is used as the light source and the other as a light detector. The LED used as light detector is reverse biased with a 5-V supply so that the photocurrent generated by the incident light discharges the capacitance. Direct digital output is provided by a simple timer circuit that measures the time taken for this discharge process from 5V (logic 1) to 1.7V (logic 0). This sensing concept has been applied in flow analysis by constructing an optical flow cell with a pair of LEDs. Calibration of the integrated optical flow cell using a dye resulted in a linear response that obeys the Beer-Lambert law. The flow rate, dynamic range, sensitivity and limits of detection were investigated. The system was also used for pH determination in the range of pH 2.5-6.8 using bromocresol green (BCG). The pK(a) of BCG was successfully determined by this technique.
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %
Sigma-Aldrich
Bromocresol Green sodium salt, ACS reagent, 90% (HPLC), Dye content 90 %