MilliporeSigma
  • Characterisation of functional domains in fission yeast Ams2 that are required for core histone gene transcription.
Characterisation of functional domains in fission yeast Ams2 that are required for core histone gene transcription.
Scientific reports (2016-12-03)
Yuko Takayama, Masaki Shirai, Fumie Masuda
ABSTRACT
Histone gene expression is regulated in a cell cycle-dependent manner, with a peak at S phase, which is crucial for cell division and genome integrity. However, the detailed mechanisms by which expression of histone genes are tightly regulated remain largely unknown. Fission yeast Ams2, a GATA-type zinc finger motif-containing factor, is required for activation of S phase-specific core histone gene transcription. Here we report the molecular characterisation of Ams2. We show that the zinc finger motif in Ams2 is necessary to bind the histone gene promoter region and to activate histone gene transcription. An N-terminal region of Ams2 acts as a self-interaction domain. Intriguingly, N-terminally truncated Ams2 binds to the histone gene promoters, but does not fully activate histone gene transcription. These observations imply that Ams2 self-interactions are required for efficient core histone gene transcription. Moreover, we show that Ams2 interacts with Teb1, which itself binds to the core histone gene promoters. We discuss the relationships between Ams2 domains and efficient transcription of the core histone genes in fission yeast.
Sigma-Aldrich
Monoclonal Anti-PSTAIR antibody produced in mouse, clone PSTAIR, ascites fluid