MilliporeSigma
  • Phagocytosis does not play a major role in naturally acquired transmission-blocking immunity to Plasmodium falciparum malaria.
Phagocytosis does not play a major role in naturally acquired transmission-blocking immunity to Plasmodium falciparum malaria.
Infection and immunity (1999-05-04)
J Healer, A Graszynski, E Riley
ABSTRACT
Phagocytosis of Plasmodium falciparum sexual stages in vitro and within the mosquito midgut was assayed in order to assess its role in transmission-blocking immunity to malaria. Both monocytes/macrophages (MM) and polymorphonuclear neutrophils (PMN) phagocytosed malarial gametes in vitro, but levels of phagocytosis were low. Intraerythrocytic gametocytes were not susceptible to phagocytosis. In vitro phagocytosis was positively correlated with levels of antibodies against the gamete surface proteins Pfs230 and Pfs48/45. Immunoglobulin G (IgG) subclass analysis revealed that phagocytosis was correlated with levels of antigamete IgG1. In vivo membrane-feeding experiments were performed in the presence of both pooled and individual malaria immune sera. The phagocytic process proceeded less efficiently in vivo than in vitro, which may be related to the lower ambient temperature (26 degrees C, compared with 37 degrees C). Finally, although we found a correlation between the ability of a serum to promote phagocytosis in vitro and the presence of antibodies against transmission-blocking target antigens, we were unable to demonstrate a role for MM- or PMN-mediated phagocytosis in reduction of infectivity of the malarial parasite to mosquitoes.
Sigma-Aldrich
Anti-Sheep Red Blood Cell Stroma antibody produced in rabbit, fractionated antiserum, lyophilized powder