MilliporeSigma
  • Inhibition of the Hexosamine Biosynthetic Pathway by targeting PGM3 causes breast cancer growth arrest and apoptosis.
Inhibition of the Hexosamine Biosynthetic Pathway by targeting PGM3 causes breast cancer growth arrest and apoptosis.
Cell death & disease (2018-03-09)
Francesca Ricciardiello, Giuseppina Votta, Roberta Palorini, Isabella Raccagni, Laura Brunelli, Alice Paiotta, Francesca Tinelli, Giuseppe D'Orazio, Silvia Valtorta, Luca De Gioia, Roberta Pastorelli, Rosa Maria Moresco, Barbara La Ferla, Ferdinando Chiaradonna
ABSTRACT
Cancer aberrant N- and O-linked protein glycosylation, frequently resulting from an augmented flux through the Hexosamine Biosynthetic Pathway (HBP), play different roles in tumor progression. However, the low specificity and toxicity of the existing HBP inhibitors prevented their use for cancer treatment. Here we report the preclinical evaluation of FR054, a novel inhibitor of the HBP enzyme PGM3, with a remarkable anti-breast cancer effect. In fact, FR054 induces in different breast cancer cells a dramatic decrease in cell proliferation and survival. In particular, in a model of Triple Negative Breast Cancer (TNBC) cells, MDA-MB-231, we show that these effects are correlated to FR054-dependent reduction of both N- and O-glycosylation level that cause also a strong reduction of cancer cell adhesion and migration. Moreover we show that impaired survival of cancer cells upon FR054 treatment is associated with the activation of the Unfolded Protein Response (UPR) and accumulation of intracellular ROS. Finally, we show that FR054 suppresses cancer growth in MDA-MB-231 xenograft mice, supporting the advantage of targeting HBP for therapeutic purpose and encouraging further investigation about the use of this small molecule as a promising compound for breast cancer therapy.
Sigma-Aldrich
Anti-Integrin beta1 Antibody, Cytosolic, serum, Chemicon®
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, clone P4G11, Chemicon®, from mouse
Sigma-Aldrich
Anti-UAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution