MilliporeSigma
  • Cellular distribution and function of ion channels involved in transport processes in rat tracheal epithelium.
Cellular distribution and function of ion channels involved in transport processes in rat tracheal epithelium.
Physiological reports (2017-06-24)
Anne Hahn, Johannes Faulhaber, Lalita Srisawang, Andreas Stortz, Johanna J Salomon, Marcus A Mall, Stephan Frings, Frank Möhrlen
ABSTRACT
Transport of water and electrolytes in airway epithelia involves chloride-selective ion channels, which are controlled either by cytosolic Ca2+ or by cAMP The contributions of the two pathways to chloride transport differ among vertebrate species. Because rats are becoming more important as animal model for cystic fibrosis, we have examined how Ca2+- dependent and cAMP- dependent Cl- secretion is organized in the rat tracheal epithelium. We examined the expression of the Ca2+-gated Cl- channel anoctamin 1 (ANO1), the cystic fibrosis transmembrane conductance regulator (CFTR) Cl- channel, the epithelial Na+ channel ENaC, and the water channel aquaporin 5 (AQP5) in rat tracheal epithelium. The contribution of ANO1 channels to nucleotide-stimulated Cl- secretion was determined using the channel blocker Ani9 in short-circuit current recordings obtained from primary cultures of rat tracheal epithelial cells in Ussing chambers. We found that ANO1, CFTR and AQP5 proteins were expressed in nonciliated cells of the tracheal epithelium, whereas ENaC was expressed in ciliated cells. Among nonciliated cells, ANO1 occurred together with CFTR and Muc5b and, in addition, in a different cell type without CFTR and Muc5b. Bioelectrical studies with the ANO1-blocker Ani9 indicated that ANO1 mediated the secretory response to the nucleotide uridine-5'-triphosphate. Our data demonstrate that, in rat tracheal epithelium, Cl- secretion and Na+ absorption are routed through different cell types, and that ANO1 channels form the molecular basis of Ca2+-dependent Cl- secretion in this tissue. These characteristic features of Cl--dependent secretion reveal similarities and distinct differences to secretory processes in human airways.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt solution, HPLC purified, aqueous solution for RNA polymerase transcription
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Sigma-Aldrich
Forskolin, For use in molecular biology applications