MilliporeSigma
  • Aging-related dysregulation in enteric dopamine and angiotensin system interactions: implications for gastrointestinal dysfunction in the elderly.
Aging-related dysregulation in enteric dopamine and angiotensin system interactions: implications for gastrointestinal dysfunction in the elderly.
Oncotarget (2018-03-16)
Pablo Garrido-Gil, Antonio Dominguez-Meijide, Rosario Moratalla, Maria J Guerra, Jose L Labandeira-Garcia
ABSTRACT
Gastrointestinal dysfunction is a common problem in the elderly. Aging-related changes in interactions between local dopaminergic and renin-angiotensin systems (RAS) have been observed in the brain, renal and vascular tissues. However, it is not known if these interactions also occur in the gut, and are dysregulated with aging. We showed a mutual regulation between the colonic dopaminergic system and RAS using young and aged mice deficient for major angiotensin and dopamine receptors. Aged rats showed a marked decrease in colonic dopamine D2 receptor expression, together with an increase in angiotensin type 1 (AT1) receptor expression, a decrease in angiotensin type 2 (AT2) receptor expression (i.e. an increase in the RAS pro-inflammatory arm activity), and increased levels of inflammatory and oxidative markers. Aged rats also showed increased levels of colonic dopamine and noradrenalin, and a marked decrease in acetylcholine and serotonin levels. The present observations contribute to explain an aging-related pro-inflammatory state and dysregulation in gastrointestinal function, which may be counteracted by treatment of aged animals with the AT1 receptor blocker candesartan.
Sigma-Aldrich
Anti-DRD1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit