MilliporeSigma
Defining transcribed regions using RNA-seq.
Nature protocols (2010-02-06)
Brian T Wilhelm, Samuel Marguerat, Ian Goodhead, Jürg Bähler
ABSTRACT
Next-generation sequencing technologies are revolutionizing genomics research. It is now possible to generate gigabase pairs of DNA sequence within a week without time-consuming cloning or massive infrastructure. This technology has recently been applied to the development of 'RNA-seq' techniques for sequencing cDNA from various organisms, with the goal of characterizing entire transcriptomes. These methods provide unprecedented resolution and depth of data, enabling simultaneous quantification of gene expression, discovery of novel transcripts and exons, and measurement of splicing efficiency. We present here a validated protocol for nonstrand-specific transcriptome sequencing via RNA-seq, describing the library preparation process and outlining the bioinformatic analysis procedure. While sample preparation and sequencing take a fairly short period of time (1-2 weeks), the downstream analysis is by far the most challenging and time-consuming aspect and can take weeks to months, depending on the experimental objectives.
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
D-Leucine, ReagentPlus®, 99%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)