MilliporeSigma
  • Isolation techniques for anthocyanidin 3,5-diglucosides and their related chemicals using supramolecules technique, and two solid-phase extraction cartridges.
Isolation techniques for anthocyanidin 3,5-diglucosides and their related chemicals using supramolecules technique, and two solid-phase extraction cartridges.
Journal of chromatography. A (2014-06-04)
Takayuki Asada, Yoriko Koi, Risa Arakawa, Fengxian Zhu, Mayumi Sadaoka, Hirotoshi Tamura
ABSTRACT
Isolation techniques for anthocyanidin 3,5-diglucosides and their derivatives from three types of supramolecular pigments were newly developed using two SPE (Discovery DPA-6S and DSC-SCX) cartridges. Initially, malonylawobanin from commelinin supramolecular complex yielded 79.5% with 85.7% purity using DPA-6S, while DSC-SCX gave a rather lower yield (62.5%) of malonylawobanin with high purity (90.8%). Delphinidin 3,5-diglucoside and awobanin from acidic hydrolysis of commelinin were easily separated using DPA-6S. Secondary, artificial Perilla supramolecules (reconstructed from Perilla crude pigment, flavocommelin and Mg(2+)) result in a high purity of a malonylshisonin and shisonin mixture (96.1%) with DSC-SCX. Finally, succinylcyanin, which is a component of protocyanin, yielded 64.3% with 86.1% purity using DPA-6S and 51.8% with 74.1% purity using DSC-SCX. Moreover, cyanidin 3,5-diglucoside yielded 43.4% with 97.0% purity using DPA-6S after partial acidic hydrolysis of protocyanin.
Product Description
Supelco
Discovery® DPA-6S SPE Tube, bed wt. 50 mg, volume 1 mL, pk of 108
Supelco
Discovery® DPA-6S SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® DPA-6S SPE Tube, bed wt. 2 g, volume 20 mL, pk of 20
Supelco
Discovery® DPA-6S SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® DPA-6S SPE Tube, bed wt. 250 mg, volume 6 mL, pk of 30