
Zolmitriptan for system suitability

Synonym(s):
Zolmitriptan, (4S)-4-((3-(2-(Dimethylamino)ethyl)-1H-indol-5-yl)methyl)-2-oxazolidinone, (S)-4-((3-(2-(Dimethylamino)ethyl)indol-5-yl)methyl)-2-oxazolidinone, 311C90
Empirical Formula (Hill Notation):
C16H21N3O2
CAS Number:
Molecular Weight:
287.36
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing