Skip to Content
MilliporeSigma
Zinc iodide ≥98%

Zinc iodide

Zinc iodide ≥98%
Synonym(s):
Diiodozinc, Zinc diiodide, Zinc iodide
Linear Formula:
ZnI2
CAS Number:
Molecular Weight:
319.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade