Skip to Content
MilliporeSigma
Yohimbine hydrochloride European Pharmacopoeia (EP) Reference Standard

Yohimbine hydrochloride

Yohimbine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride, Yohimbine hydrochloride
Empirical Formula (Hill Notation):
C21H26N2O3 · HCl
CAS Number:
Molecular Weight:
390.90

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade