Skip to Content
MilliporeSigma
Xanthone pharmaceutical secondary standard, certified reference material

Xanthone

Xanthone pharmaceutical secondary standard, certified reference material
Synonym(s):
9-Xanthenone, Xanthone
Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade