Skip to Content
MilliporeSigma
Uridine 5′-diphospho-N-acetylgalactosamine disodium salt ≥97%

Uridine 5′-diphospho-N-acetylgalactosamine disodium salt

Uridine 5′-diphospho-N-acetylgalactosamine disodium salt ≥97%
Synonym(s):
UDP-GalNAc, UDP-N-acetylgalactosamine, Uridine[5′]diphospho[1](2-acetamino-2-deoxy-α-D-galactopyranose) disodium salt
Empirical Formula (Hill Notation):
C17H25N3Na2O17P2
CAS Number:
Molecular Weight:
651.32

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing