
Uric acid

Synonym(s):
2,6,8-Trihydroxypurine
Empirical Formula (Hill Notation):
C5H4N4O3
CAS Number:
Molecular Weight:
168.11
Products
Brand
Biological Source
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing