Skip to Content
MilliporeSigma
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide ≥97% (HPLC), powder

(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide

(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide ≥97% (HPLC), powder
Synonym(s):
CCK-8, Cholecystokinin octapeptide, Pancreozymin C-terminal octapeptide, Sincalide
Empirical Formula (Hill Notation):
C49H62N10O16S3
CAS Number:
Molecular Weight:
1143.27
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing