Skip to Content
MilliporeSigma
Tungstosilicic acid hydrate

Tungstosilicic acid hydrate

Tungstosilicic acid hydrate
Synonym(s):
Silicotungstic acid, Tungstosilicic acid hydrate
Linear Formula:
H4[Si(W3O10)4] · x H2O
CAS Number:
Molecular Weight:
2878.17 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Special Grade