Skip to Content
MilliporeSigma
Tubocurarine hydrochloride pentahydrate ≥97%

Tubocurarine hydrochloride pentahydrate

Tubocurarine hydrochloride pentahydrate ≥97%
Synonym(s):
(+)-Tubocurarine hydrochloride
Empirical Formula (Hill Notation):
C37H41ClN2O6 · HCl · 5H2O
CAS Number:
Molecular Weight:
771.72

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing