
TSKgel® Octyl-80Ts (5 µm) HPLC Columns

Synonym(s):
TSKgel® Reversed Phase HPLC Column, TSKgel® Reversed Phase HPLC Guard Cartridge
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing