Skip to Content
MilliporeSigma
Tris(trimethylsilyl)phosphine 95%

Tris(trimethylsilyl)phosphine

Tris(trimethylsilyl)phosphine 95%
Linear Formula:
[(CH3)3Si]3P
CAS Number:
Molecular Weight:
250.54

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing