Skip to Content
MilliporeSigma
Tris(4-formylphenyl)amine 97%

Tris(4-formylphenyl)amine

Tris(4-formylphenyl)amine 97%
Synonym(s):
4,4′,4″-Nitrilotrisbenzaldehyde, 4,4′,4″-Triformyltriphenylamine
Empirical Formula (Hill Notation):
C21H15NO3
CAS Number:
Molecular Weight:
329.35

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing