Skip to Content
MilliporeSigma
Triphenylmethanol United States Pharmacopeia (USP) Reference Standard

Triphenylmethanol

Triphenylmethanol United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Triphenylcarbinol, Trityl alcohol, Triphenylmethanol
Linear Formula:
(C6H5)3COH
CAS Number:
Molecular Weight:
260.33

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3