Skip to Content
MilliporeSigma
Trazodone Related Compound F certified reference material, pharmaceutical secondary standard

Trazodone Related Compound F

Trazodone Related Compound F certified reference material, pharmaceutical secondary standard
Synonym(s):
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine monohydrochloride
Empirical Formula (Hill Notation):
C13H18Cl2N2 · HCl
CAS Number:
Molecular Weight:
309.66

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade