Skip to Content
MilliporeSigma
trans,trans-1,4-Diphenyl-1,3-butadiene 98%

trans,trans-1,4-Diphenyl-1,3-butadiene

trans,trans-1,4-Diphenyl-1,3-butadiene 98%
Synonym(s):
β,β′-Bistyryl, DPB
Linear Formula:
C6H5CH=CHCH=CHC6H5
CAS Number:
Molecular Weight:
206.28

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing