
Transplatin

Synonym(s):
trans-Platinum(II)diammine dichloride, trans-Diamminedichloroplatinum(II), trans-Dichlorodiammineplatinum(II)
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3