Skip to Content
MilliporeSigma
trans-Cinnamic acid analytical standard

trans-Cinnamic acid

trans-Cinnamic acid analytical standard
Synonym(s):
trans-3-Phenylacrylic acid, Cinnamic acid, (2E)-3-Phenyl-2-propenoic acid
Linear Formula:
C6H5CH=CHCOOH
CAS Number:
Molecular Weight:
148.16
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique