Skip to Content
MilliporeSigma
trans-4-Methoxy-3-buten-2-one technical grade, 90%

trans-4-Methoxy-3-buten-2-one

trans-4-Methoxy-3-buten-2-one technical grade, 90%
Linear Formula:
CH3OCH=CHCOCH3
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing