Skip to Content
MilliporeSigma
trans-3-Indoleacrylic acid 98%

trans-3-Indoleacrylic acid

trans-3-Indoleacrylic acid 98%
Empirical Formula (Hill Notation):
C11H9NO2
CAS Number:
Molecular Weight:
187.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing